3-Methylthiophene-2-carbonyl chloride ≥98%

Supplier: Matrix Scientific

012539-5G 012539-25G
101780-114EA 63.08 USD
101780-114 101780-116
3-Methylthiophene-2-carbonyl chloride ≥98%
3-Methylthiophene-2-carbonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₆H₅ClOS
MW: 160.62 g/mol
Boiling Pt: 216…220 °C
MDL Number: MFCD00130091
CAS Number: 61341-26-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 216-220°
ND20 1.582

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR