Tetrafluoroterephthalonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: Perfluoroterephthalonitrile

011293-25G 011293-100G
101780-094EA 75.38 USD
101780-094 101780-096
Tetrafluoroterephthalonitrile ≥98%
Tetrafluoroterephthalonitrile

Hazard: Toxic/Irritant

Formula: C₈F₄N₂
MW: 200.1 g/mol
Melting Pt: 197-199°C
MDL Number: MFCD00001776
CAS Number: 1835-49-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 197-199°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR