5-Chloro-2-fluoropyridine ≥98%

Supplier: Matrix Scientific

011273-5G 011273-100G 011273-25G
101780-076EA 29.23 USD
101780-076 101780-080 101780-078
5-Chloro-2-fluoropyridine ≥98%
5-Chloro-2-fluoropyridine

Purity: 98% CAS: 1480-65-5 MDL Number: MFCD04112513 Molecular Formula: C5H3ClFN Molecular Weight: 131,54 Hazard: Flammable/Toxic

Formula: C₅H₃ClFN
MDL Number: MFCD04112513
CAS Number: 1480-65-5
UN: 1992
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR