2-Chloroisonicotinonitrile ≥98%

Supplier: Matrix Scientific

011230-25G 011230-100G
101780-068EA 23.08 USD
101780-068 101780-070
2-Chloroisonicotinonitrile ≥98%
2-Chloroisonicotinonitrile

Hazard: Toxic/Irritant

Formula: C₆H₃ClN₂
Boiling Pt: 104…106 °C (15 mmHg)
Melting Pt: 49 °C
MDL Number: MFCD00174318
CAS Number: 33252-30-1
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 104-106°/1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR