3-Amino-2-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Methylpyridin-3-amine, 2-Methyl-3-pyridylamine, 3-Amino-2-picoline

011185-5G 011185-25G
101780-054EA 23.08 USD
101780-054 101780-056
3-Amino-2-methylpyridine ≥98%
3-Amino-2-methylpyridine

Hazard: Toxic

Formula: C₆H₈N₂
MW: 108.14 g/mol
Melting Pt: 115…118 °C
MDL Number: MFCD03788195
CAS Number: 3430-10-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 115-118°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR