3,4-Diaminopyridine ≥98%
Supplier: Matrix Scientific
011183-25G
011183-5G
101780-052EA
90.77
USD
101780-052
101780-050
3,4-Diaminopyridine ≥98%
3,4-Diaminopyridine
Hazard: Toxic/Irritant
Formula:
C₅H₇N₃ Melting Pt: 218…220 °C |
MDL Number:
MFCD00006401 CAS Number: 54-96-6 UN: 2811 ADR: 6.1,II |
Specification Test Results
Melting point | 218-220° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...