2,6-Dichloro-3-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

011155-5G 011155-25G
101780-046EA 73.85 USD
101780-046 102832-830
2,6-Dichloro-3-(trifluoromethyl)pyridine ≥95%
2,6-Dichloro-3-(trifluoromethyl)pyridine

Hazard: Toxic/Irritant

Formula: C₆H₂Cl₂F₃N
MW: 215.99 g/mol
Boiling Pt: 194-196°C
Melting Pt: 6-7°C
MDL Number: MFCD00042245
CAS Number: 55304-75-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 194-196°
ND20 1.4855

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR