α-Chloro-2,5-difluorotoulene ≥97%

Supplier: Matrix Scientific

011148-5G 011148-25G
101780-040EA 56.92 USD
101780-040 101780-042
α-Chloro-2,5-difluorotoulene ≥97%
α-Chloro-2,5-difluorotoulene

Hazard: Corrosive/Irritant

Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 62…64 °C (14 Torr)
MDL Number: MFCD01090989
CAS Number: 495-07-8
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR