α-Chloro-2,4-difluorotoulene ≥98%

Supplier: Matrix Scientific

011147-25G 011147-100G
101780-036EA 346.15 USD
101780-036 101780-038
α-Chloro-2,4-difluorotoulene ≥98%
α-Chloro-2,4-difluorotoulene

Hazard: Corrosive/Irritant

Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 60-62°C/14 Torr
MDL Number: MFCD01090988
CAS Number: 452-07-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR