5-Bromopyridine-2-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromopicolinonitrile, 5-Bromo-2-cyanopyridine
011142-100G
011142-25G
101780-030EA
181.54
USD
101780-030
101780-028
5-Bromopyridine-2-carbonitrile ≥95%
5-Bromopyridine-2-carbonitrile
Hazard: Toxic/Irritant
Formula:
C₆H₃BrN₂ MW: 183.01 Boiling Pt: 100 °C (3 mmHg) Melting Pt: 128…132 °C |
MDL Number:
MFCD00234144 CAS Number: 97483-77-7 UN: 2811 ADR: 6.1 III |
Specification Test Results
Boiling point | 100°/3mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...