3-Fluoro-4-methoxybenzoyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3-fluoro-4-methoxybenzoylchloride, 3-Fluoro-4-methoxybenzene-1-carbonyl chloride
011127-5G
011127-25G
101780-022EA
20
USD
101780-022
101780-024
3-Fluoro-4-methoxybenzoyl chloride ≥97%
3-Fluoro-4-methoxybenzoyl chloride
Hazard: Corrosive/Irritant
Formula:
C₈H₆ClFO₂ MW: 188.59 g/mol Boiling Pt: 135 °C (11 Torr) Melting Pt: 69 °C |
MDL Number:
MFCD03428589 CAS Number: 3907-15-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...