2,4-Dichloro-6-methylpyrimidine ≥99%
Supplier: Matrix Scientific
011105-5G
011105-100G
101780-010EA
16.92
USD
101780-010
101780-014
2,4-Dichloro-6-methylpyrimidine ≥99%
2,4-Dichloro-6-methylpyrimidine
TCA listed: T
Hazard: Corrosive
Formula:
C₅H₄Cl₂N₂ MW: 163.01 g/mol Boiling Pt: 218…220 °C Melting Pt: 44…47 °C |
MDL Number:
MFCD00006064 CAS Number: 5424-21-5 UN: 3263 ADR: 8,II |
Specification Test Results
Boiling point | 218-220° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...