3-Aminopyridine ≥99%
Supplier: Matrix Scientific
Synonyms:
Pyridin-3-amine, 3-Pyridylamine
011103-500G
011103-100G
101780-008EA
178.46
USD
101780-008
101780-006
3-Aminopyridine ≥99%
3-Aminopyridine
TCA listed: T
Hazard: Toxic/Air Sensitive/Hygroscopic
Formula:
C₅H₆N₂ Boiling Pt: 104…106 °C (20 mmHg) Melting Pt: 59…63 °C |
MDL Number:
MFCD00006400 CAS Number: 462-08-8 UN: 2671 ADR: 6.1,II |
Specification Test Results
Boiling point | 104-106°/2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...