4-Amino-3-(trifluoromethyl)benzonitrile ≥98%
Supplier: Matrix Scientific
011083-5G
011083-25G
011083-100G
101780-000EA
21.54
USD
101780-000
101780-002
101780-004
4-Amino-3-(trifluoromethyl)benzonitrile ≥98%
4-Amino-3-(trifluoromethyl)benzonitrile
Hazard: Toxic
Formula:
C₈H₅F₃N₂ Boiling Pt: 100 °C (0.1 mmHg) Melting Pt: 64…65 °C |
MDL Number:
MFCD00275473 CAS Number: 327-74-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...