2,5-Bis(trifluoromethyl)benzyl bromide

Supplier: Matrix Scientific

Synonyms: 1-(Bromomethyl)-2,5-bis(trifluoromethyl)benzene, 2-(Bromomethyl)-1,4-bis(trifluoromethyl)benzene

009726-5G 009726-1G
101779-992EA 95.38 USD
101779-992 101779-990
2,5-Bis(trifluoromethyl)benzyl bromide
2,5-Bis(trifluoromethyl)benzyl bromide

Hazard: Corrosive/Lachrymator

Formula: C₉H₅BrF₆
MW: 307.03 g/mol
MDL Number: MFCD00799485
CAS Number: 302911-98-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR