α-Bromo-4-chloro-2-fluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-fluorobenzyl bromide, 1-(Bromomethyl)-4-chloro-2-fluorobenzene

009619-25G 009619-100G 009619-5G
101779-982EA 67.69 USD
101779-982 101779-984 101779-980
α-Bromo-4-chloro-2-fluorotoluene ≥98%
α-Bromo-4-chloro-2-fluorotoluene

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₅BrClF
MW: 223.47
MDL Number: MFCD00831105
CAS Number: 71916-82-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR