4-Fluoro-o-toluonitrile ≥97%

Supplier: Matrix Scientific

009518-5G 009518-25G
101779-974EA 15.38 USD
101779-974 101779-976
4-Fluoro-o-toluonitrile ≥97%
4-Fluoro-o-toluonitrile

Hazard: Toxic

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 70…74 °C
MDL Number: MFCD03095106
CAS Number: 147754-12-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 70-74°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR