α-Bromo-4-chloro-3-fluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 4-(Bromomethyl)-1-chloro-2-fluorobenzene, 4-Chloro-3-fluorobenzyl bromide

009486-25G 009486-5G
101779-972EA 47.69 USD
101779-972 102832-808
α-Bromo-4-chloro-3-fluorotoluene ≥98%
α-Bromo-4-chloro-3-fluorotoluene

Hazard: Corrosive/Refrigerate/Lachrymator

Formula: C₇H₅BrClF
MW: 223.47 g/mol
MDL Number: MFCD04115859
CAS Number: 206362-80-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR