2-Bromo-2'-(trifluoromethoxy)acetophenone

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethoxy)phenacyl bromide

009250-5G 009250-1G
101779-956EA 1000 USD
101779-956 101779-954
2-Bromo-2'-(trifluoromethoxy)acetophenone
2-Bromo-2'-(trifluoromethoxy)acetophenone

Hazard: Corrosive/Refrigerate

Formula: C₉H₆BrF₃O₂
MDL Number: MFCD03094424
CAS Number: 530141-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR