2,4,6-Trifluoropyridine ≥98%
Supplier: Matrix Scientific
009238-5G
009238-25G
101779-950EA
238.46
USD
101779-950
101779-952
2,4,6-Trifluoropyridine ≥98%
2,4,6-Trifluoropyridine
Hazard: Flammable
Formula:
C₅H₂F₃N Boiling Pt: 94°C Density: 1.499 |
MDL Number:
MFCD03001161 CAS Number: 3512-17-2 UN: 2924 ADR: 3,II |
Specification Test Results
Boiling point | 96.5-97° |
Density | 1.499 |
ND20 | 1.412 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...