3-Butyne-1-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

009231-25G 009231-100G
101779-946EA 35.38 USD
101779-946 101779-948
3-Butyne-1-ol ≥97%
3-Butyne-1-ol

Hazard: Flammable

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 129…130 °C
Melting Pt: –63 °C
Density: 0.9236/20°C
MDL Number: MFCD00002955
CAS Number: 927-74-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Boiling point 129-130°
Density 0.927
ND20 1.441

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR