Thiazol-2-ylmethanamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(Aminomethyl)-1,3-thiazole hydrochloride, (1,3-Thiazol-2-yl)methylamine hydrochloride, 2-Aminomethylthiazole hydrochloride

009227-250MG 009227-1G
101779-942EA 106.15 USD
101779-942 101779-944
Thiazol-2-ylmethanamine hydrochloride
Thiazol-2-ylmethanamine hydrochloride

Hazard: Corrosive/Irritant/Hygroscopic

Formula: C₄H₇ClN₂S
MW: 150.63 g/mol
Melting Pt: 186…187 °C
MDL Number: MFCD04113607
CAS Number: 850852-85-6

Order Now

Specification Test Results

Boiling point 39°/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR