2-Vinylhexafluoroisopropanol ≥99%

Supplier: Matrix Scientific

Synonyms: 1,1,1-Trifluoro-2-(trifluoromethyl)but-3-en-2-ol

009224-5G 009224-25G
101779-938EA 653.85 USD
101779-938 101779-940
2-Vinylhexafluoroisopropanol ≥99%
2-Vinylhexafluoroisopropanol

Hazard: Corrosive

Formula: C₅H₄F₆O
MW: 194.08 g/mol
Boiling Pt: 72 °C
MDL Number: MFCD04038371
CAS Number: 19701-19-0

Order Now

Specification Test Results

Boiling point 73-74°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR