4,5,5-Trifluoropent-4-enoic acid ≥97%
Supplier: Matrix Scientific
009215-1G
009215-5G
101779-926EA
200
USD
101779-926
101779-928
4,5,5-Trifluoropent-4-enoic acid ≥97%
4,5,5-Trifluoropent-4-enoic acid
Hazard: Corrosive
Formula:
C₅H₅F₃O₂ |
MDL Number:
MFCD00156072 CAS Number: 110003-22-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...