2,3,6-Trifluoropyridine ≥99%

Supplier: Matrix Scientific

009191-1G 009191-5G
101779-908EA 52.31 USD
101779-908 101779-910
2,3,6-Trifluoropyridine ≥99%
2,3,6-Trifluoropyridine

Hazard: Flammable/Corrosive

Formula: C₅H₂F₃N
MW: 133.07 g/mol
Boiling Pt: 118-119°C
Density: 1.499
MDL Number: MFCD03001158
CAS Number: 3512-18-3
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 118-119°
Density 1.499
ND20 1.42

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR