2,2,2,3'-Tetrafluoroacetophenone ≥99%

Supplier: Matrix Scientific

Synonyms: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethanone

009161-5G 009161-1G
101779-894EA 118.46 USD
101779-894 101779-892
2,2,2,3'-Tetrafluoroacetophenone ≥99%
2,2,2,3'-Tetrafluoroacetophenone

TCA listed: T
Hazard: Flammable

Formula: C₈H₄F₄O
MW: 192.11 g/mol
Boiling Pt: 79 °C (50 mmHg)
Density: 1.372
MDL Number: MFCD00039236
CAS Number: 708-64-5

Order Now

Specification Test Results

Boiling point 79°/50mm
Density 1.372
ND20 1.4503

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR