Perfluoro-3,6,9-trioxatridecanoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2,2-Difluoro-2-(1,1,2,2-tetrafluoro-2-(1,1,2,2-tetrafluoro-2-(perfluorobutoxy)ethoxy)ethoxy)acetic acid

009139-5G 009139-25G
101779-888EA 230.77 USD
101779-888 101779-890
Perfluoro-3,6,9-trioxatridecanoic acid ≥98%
Perfluoro-3,6,9-trioxatridecanoic acid

Hazard: Corrosive

Formula: C₁₀HF₁₉O₅
MW: 562.09 g/mol
MDL Number: MFCD03094135
CAS Number: 330562-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR