Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥97%

Supplier: Matrix Scientific

Synonyms: (Chloro)(perfluorophenyl)dimethylsilane

009081-1G 009081-5G
101779-868EA 56.92 USD
101779-868 101779-870
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥97%
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride)

Hazard: Flammable/Corrosive

Formula: C₈H₆ClF₅Si
MW: 260.67 g/mol
Boiling Pt: 88…90 °C (10 mmHg)
Density: 1.367
MDL Number: MFCD00000500
CAS Number: 20082-71-7
UN: 2987
ADR: 8 II

Order Now

Specification Test Results

Boiling point 88-90°/10m
Density 1.384
ND20 1.447


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR