Perfluorobutane-1-sulfonic acid
Supplier: Matrix Scientific
Synonyms:
Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid
009050-5G
009050-25G
101779-852EA
158.46
USD
101779-852
101779-854
Perfluorobutane-1-sulfonic acid
Perfluorobutane-1-sulphonic acid
Hazard: Corrosive
Formula:
C₄HF₉O₃S Boiling Pt: 211°C |
MDL Number:
MFCD01320794 CAS Number: 375-73-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...