Methyl-3,3,3-trifluoropropanate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 3,3,3-Trifluoropropionate, 3,3,3-Trifluoropropionic acid methyll ester
009042-5G
009042-25G
101779-848EA
132.31
USD
101779-848
101779-850
Methyl-3,3,3-trifluoropropanate ≥97%
Methyl-3,3,3-trifluoropropanate
Hazard: Flammable/Irritant
Formula:
C₄H₅F₃O₂ MW: 142.08 g/mol Boiling Pt: 95-95.5°C Density: 1.3071 |
MDL Number:
MFCD02262206 CAS Number: 18830-44-9 |
Specification Test Results
Boiling point | 95-95.5° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...