Methyl-2-(trifluoromethyl)acrylate ≥97% inhibited
Supplier: Matrix Scientific
009039-1G
009039-5G
101779-842EA
138.46
USD
101779-842
101779-844
Methyl-2-(trifluoromethyl)acrylate ≥97% inhibited
Methyl-2-(trifluoromethyl)acrylate
Hazard: Flammable/Refrigerate
Formula:
C₅H₅F₃O₂ Boiling Pt: 104…105 °C |
MDL Number:
MFCD00068659 CAS Number: 382-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...