Methyl-4,4,4-trifluorobutyrate ≥97%
Supplier: Matrix Scientific
009038-1G
009038-5G
101779-838EA
87.69
USD
101779-838
101779-840
Methyl-4,4,4-trifluorobutyrate ≥97%
Methyl-4,4,4-trifluorobutyrate
Hazard: Flammable/Irritant
Formula:
C₅H₇F₃O₂ Boiling Pt: 113 °C |
CAS Number:
2365-82-4 |
Specification Test Results
Boiling point | 113° |
ND20 | 1.345 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...