Z-1-Methoxy-3,3,3-trifluoropropene ≥97%

Supplier: Matrix Scientific

009023-1G 009023-5G
101779-828EA 206.15 USD
101779-828 101779-830
Z-1-Methoxy-3,3,3-trifluoropropene ≥97%
Z-1-Methoxy-3,3,3-trifluoropropene

Hazard: Flammable

Formula: C₄H₅F₃O
Boiling Pt: 102…104 °C
MDL Number: MFCD04038317
CAS Number: 26885-67-6

Order Now

Specification Test Results

Boiling point 102-104°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR