Lithium bis(trifluoromethylsulphonyl)imide

Supplier: Matrix Scientific

Synonyms: Lithium bis(trifluoromethanesulphonimide), Lithium bis((trifluoromethyl)sulphonyl)amide, Lithium Bis(Trifluoromethanesulphonyl)Imide, Bis(trifluoromethane)sulphonimide lithium salt

009016-25G 009016-100G
101779-818EA 115.38 USD
101779-818 101779-820
Lithium bis(trifluoromethylsulphonyl)imide
Lithium bis(trifluoromethylsulphonyl)imide

TCA listed: T
Hazard: Toxic/Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₂F₆LiNO₄S₂
MW: 287.09 g/mol
Melting Pt: 238 °C
Density: 2.27
MDL Number: MFCD00210017
CAS Number: 90076-65-6
UN: 2923
ADR: 8 II

Order Now

Specification Test Results

Melting point 226°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR