Heptafluoroisopropyl acrylate ≥98%

Supplier: Matrix Scientific

008986-1G 008986-5G
101779-814EA 78.46 USD
101779-814 101779-816
Heptafluoroisopropyl acrylate ≥98%
Heptafluoroisopropyl acrylate

Hazard: Flammable/Refrigerate

Formula: C₆H₃F₇O₂
Boiling Pt: 86…87 °C
Density: 1.4
MDL Number: MFCD00274356
CAS Number: 13057-08-4

Order Now

Specification Test Results

Boiling point 86-87°
Density 1.4
ND20 1.312


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR