2-Fluoro-6-methoxybenzonitrile ≥97%

Supplier: Matrix Scientific

008957-25G 008957-100G
101779-806EA 24.62 USD
101779-806 101779-808
2-Fluoro-6-methoxybenzonitrile ≥97%
2-Fluoro-6-methoxybenzonitrile

Hazard: Toxic/Irritant

Formula: C₈H₆FNO
MW: 151.14 g/mol
Melting Pt: 102-104°C
MDL Number: MFCD00042291
CAS Number: 94088-46-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 102-104°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR