2,3,5-Trifluoro-4-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

008755-250MG 008755-1G
101779-744EA 63.08 USD
101779-744 101779-746
2,3,5-Trifluoro-4-(trifluoromethyl)pyridine ≥98%
2,3,5-Trifluoro-4-(trifluoromethyl)pyridine

Hazard: Flammable

Formula: C₆HF₆N
MW: 201.07 g/mol
Boiling Pt: 102…104 °C
CAS Number: 675602-93-4

Order Now

Specification Test Results

Boiling point 102-104°
ND20 1.3823

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR