6-Fluoro-m-toluonitrile ≥98%

Supplier: Matrix Scientific

008744-5G 008744-25G
101779-736EA 96.92 USD
101779-736 101779-738
6-Fluoro-m-toluonitrile ≥98%
6-Fluoro-m-toluonitrile

Hazard: Toxic/Irritant

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD00134542
CAS Number: 64113-84-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR