4-Methyl-3-nitrobenzenesulfonyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Methyl-3-nitrobenzene-1-sulphonyl chloride

008693-5G 008693-25G
101779-714EA 44.62 USD
101779-714 101779-716
4-Methyl-3-nitrobenzenesulfonyl chloride ≥98%
4-Methyl-3-nitrobenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₆ClNO₄S
MW: 235.65 g/mol
Boiling Pt: 142 °C (0.8 mmHg)
Melting Pt: 33…36 °C
MDL Number: MFCD00129811
CAS Number: 616-83-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Boiling point 142°/0.8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR