4-(Trifluoromethyl)thiobenzamide ≥97%

Supplier: Matrix Scientific

007667-5G 007667-25G
101779-706EA 15.38 USD
101779-706 101779-708
4-(Trifluoromethyl)thiobenzamide ≥97%
4-(Trifluoromethyl)thiobenzamide

Hazard: Toxic

Formula: C₈H₆F₃NS
MW: 205.2 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 134-136°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR