(±)-3-Buten-2-ol ≥97%
Supplier: Matrix Scientific
Synonyms:
(±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol
007660-25G
007660-100G
101779-702EA
107.69
USD
101779-702
101779-704
(±)-3-Buten-2-ol ≥97%
(±)-3-Buten-2-ol
Hazard: Flammable/Toxic
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 96-98°C Density: 0.838 Flash Pt: 20°C |
MDL Number:
MFCD00004543 CAS Number: 598-32-3 UN: 1986 ADR: 3,II |
Specification Test Results
Boiling point | 96-98° |
Density | 0.838 |
ND20 | 1.414 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...