1-(3-Aminopropyl)pyrrolidine ≥97%

Supplier: Matrix Scientific

007649-25G 007649-5G
101779-688EA 267.69 USD
101779-688 101779-686
1-(3-Aminopropyl)pyrrolidine ≥97%
1-(3-Aminopropyl)pyrrolidine

Hazard: Corrosive/Stored under Argon

Formula: C₇H₁₆N₂
Boiling Pt: 78°C/ 15mm
MDL Number: MFCD00014100
CAS Number: 23159-07-1

Order Now

Specification Test Results

Boiling point 78°/ 15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR