N-Methyldipropylamine ≥98%

Supplier: Matrix Scientific

007624-25G 007624-5G
101779-676EA 390.77 USD
101779-676 101779-674
N-Methyldipropylamine ≥98%
N-Methyldipropylamine

Hazard: Corrosive

Formula: C₇H₁₇N
Boiling Pt: 117 °C
MDL Number: MFCD00048683
CAS Number: 3405-42-3

Order Now

Specification Test Results

Boiling point 117°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR