N-Methyldi-N-butylamine ≥98%

Supplier: Matrix Scientific

007621-5G 007621-25G
101779-670EA 73.85 USD
101779-670 101779-672
N-Methyldi-N-butylamine ≥98%
N-Methyldi-N-butylamine

TCA listed: T
Hazard: Flammable/Corrosive

Formula: C₉H₂₁N
Boiling Pt: 49…51 °C (10 mmHg)
Density: 0.745
MDL Number: MFCD00009430
CAS Number: 3405-45-6

Order Now

Specification Test Results

Boiling point 49-51°/10m
Density 0.745
ND20 1.4178


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR