3-Methoxy-1-propanol ≥98%

Supplier: Matrix Scientific

Synonyms: Trimethylene glycol monomethyl ether

007618-25G 007618-100G
101779-666EA 21.54 USD
101779-666 101779-668
3-Methoxy-1-propanol ≥98%
3-Methoxy-1-propanol

Hazard: Flammable

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 151°C
Density: 0.95
MDL Number: MFCD00039550
CAS Number: 1589-49-7
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Boiling point 151°
Density 0.95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR