1-(Dimethylamino)isopropylamine ≥97%

Supplier: Matrix Scientific

Synonyms: N1,N1-Dimethyl-1,2-propanediamine dihydrochloride

007576-1G 007576-5G
101779-638EA 69.23 USD
101779-638 101779-640
1-(Dimethylamino)isopropylamine ≥97%
1-(Dimethylamino)isopropylamine

Hazard: Corrosive

Formula: C₅H₁₄N₂
MW: 102.18 g/mol
Boiling Pt: 113…115 °C
MDL Number: MFCD00014812
CAS Number: 108-15-6

Order Now

Specification Test Results

Boiling point 113°-115°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR