4-(Dimethylamino)butylamine ≥97%

Supplier: Matrix Scientific

007572-1G 007572-5G
101779-632EA 150.77 USD
101779-632 101779-634
4-(Dimethylamino)butylamine ≥97%
4-(Dimethylamino)butylamine

Hazard: Corrosive

Formula: C₆H₁₆N₂
Boiling Pt: 73 °C ( 35 mmHg)
MDL Number: MFCD00047977
CAS Number: 3529-10-0

Order Now

Specification Test Results

Boiling point 73°/ 35mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR