4-(Diethylamino)butylamine ≥97%

Supplier: Matrix Scientific

Synonyms: N1,N1-Diethylbutane-1,4-diamine

007561-1G 007561-5G
101779-620EA 36.92 USD
101779-620 101779-622
4-(Diethylamino)butylamine ≥97%
4-(Diethylamino)butylamine

Hazard: Corrosive/Air Sensitive/Moisture Sensitive

Formula: C₈H₂₀N₂
MW: 144.26 g/mol
Boiling Pt: 55°C/ 3mm
Density: 0.83
MDL Number: MFCD00014836
CAS Number: 27431-62-5

Order Now

Specification Test Results

Boiling point 55°/ 3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR