2-(Chloromethyl)naphthalene ≥90%

Supplier: Matrix Scientific

007537-1G 007537-5G
101779-594EA 38.46 USD
101779-594 101779-596
2-(Chloromethyl)naphthalene ≥90%
2-(Chloromethyl)naphthalene

Hazard: Corrosive/Irritant-Harmful/Lachrymator

Formula: C₁₁H₉Cl
MW: 176.65 g/mol
Boiling Pt: 135°C/3mm
Melting Pt: 45°C
MDL Number: MFCD00046488
CAS Number: 2506-41-4
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Boiling point 135°/3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR