4-Ethylbenzylamine ≥97%

Supplier: Matrix Scientific

007464-5G 007464-1G
101779-560EA 296.92 USD
101779-560 101779-558
4-Ethylbenzylamine ≥97%
4-Ethylbenzylamine

Hazard: Corrosive/Air Sensitive

Formula: C₉H₁₃N
Boiling Pt: 110°C/8mm
Density: 0.94
MDL Number: MFCD01529871
CAS Number: 7441-43-2

Order Now

Specification Test Results

Boiling point 110°/8mm
Density 0.94


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR